siRNA Details
| Gene Symbol: Il34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025766.1 | NP_001020937.1 | 5, 6 | 909 | ||
| XM_006255599.2 | XP_006255661.1 | 6, 7 | 1275 | |||
| XM_006255600.2 | XP_006255662.1 | 6, 7 | 1272 | |||
| XM_008772527.1 | XP_008770749.1 | 6, 7 | 980 | |||
| GenBank mRNA | BC076391.1 | - | 6, 7 | 1038 | ||
| BC098808.1 | - | 5, 6 | 909 | 7376793 | ||

Il34
Genomic Map