siRNA Details
| Gene Symbol: Zfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255656.2 | XP_006255718.1 | 4 | 500 | ||
| XM_006255657.2 | XP_006255719.1 | 4 | 1258 | |||
| XM_006255660.2 | XP_006255722.1 | 4 | 1224 | |||
| XM_006255662.2 | XP_006255724.1 | 4 | 451 | |||
| XM_006255663.2 | XP_006255725.1 | 3 | 1155 | |||
| XM_008772583.1 | XP_008770805.1 | 5 | 1455 | |||
| XM_008772584.1 | XP_008770806.1 | 5 | 1452 | |||
| XM_008772585.1 | XP_008770807.1 | 5 | 1404 | |||
| XM_008772586.1 | XP_008770808.1 | 4 | 828 | |||
| XM_574241.6 | XP_574241.4 | 4 | 467 | |||

Zfp1
Genomic Map