siRNA Details
| Gene Symbol: Klhl36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017511.1 | NP_001017511.1 | 2 | 158 | ||
| XM_006255718.2 | XP_006255780.1 | 2 | 270 | |||
| XM_008772641.1 | XP_008770863.1 | 2 | 1520 | |||
| GenBank mRNA | BC081914.1 | - | 2 | 158 | 7112173 | |

Klhl36
Genomic Map