siRNA Details
| Gene Symbol: RGD1559896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109134.1 | NP_001102604.1 | 3 | 565 | ||
| XM_008772642.1 | XP_008770864.1 | 3 | 582 | |||
| XM_008772643.1 | XP_008770865.1 | 3 | 465 | |||
| GenBank mRNA | FQ212098.1 | - | 3 | 307 | ||

RGD1559896
Genomic Map