siRNA Details
| Gene Symbol: Ncoa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227758.2 | XP_006227820.1 | 9 | 1861 | ||
| XM_008758675.1 | XP_008756897.1 | 9 | 1828 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncoa7| Gene Symbol: Ncoa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227758.2 | XP_006227820.1 | 9 | 1861 | ||
| XM_008758675.1 | XP_008756897.1 | 9 | 1828 | |||