siRNA Details
| Gene Symbol: Rspo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100990.1 | NP_001094460.1 | 2 | 372 | ||
| GenBank mRNA | EU030376.1 | - | 2 | 372 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rspo3| Gene Symbol: Rspo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100990.1 | NP_001094460.1 | 2 | 372 | ||
| GenBank mRNA | EU030376.1 | - | 2 | 372 | ||