siRNA Details
Gene Symbol: Zdhhc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039342.2 | NP_001034431.2 | 3 | 309 | ||
XM_006227796.2 | XP_006227858.1 | 2 | 270 | |||
XM_006227797.2 | XP_006227859.1 | 2 | 270 | |||
GenBank mRNA | AY886527.1 | - | 3 | 341 |
