siRNA Details
| Gene Symbol: Wtap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113542.1 | NP_001107014.1 | 8 | 860 | ||
| XM_006227892.2 | XP_006227954.1 | 8 | 840 | |||
| XM_006227893.2 | XP_006227955.1 | 8 | 1018 | |||
| GenBank mRNA | CO561762.1 | - | 2 | 256 | ||
| DV714133.1 | - | 2 | 83 | |||
| FQ212824.1 | - | 1 | 67 | |||

Wtap
Genomic Map