siRNA Details
| Gene Symbol: Wtap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113542.1 | NP_001107014.1 | 6 | 517 | ||
| NM_001113543.1 | NP_001107015.1 | 6 | 517 | |||
| NM_001113544.1 | NP_001107016.1 | 6 | 451 | |||
| XM_006227892.2 | XP_006227954.1 | 6 | 497 | |||
| XM_006227893.2 | XP_006227955.1 | 6 | 675 | |||
| GenBank mRNA | CB613741.1 | - | 6 | 446 | ||
| CB613959.1 | - | 6 | 451 | |||
| CK478322.1 | - | 6 | 517 | |||
| CV108433.1 | - | 6 | 514 | |||
| CX570823.1 | - | 2 | 163 | |||
| FQ228682.1 | - | 6 | 556 | |||

Wtap
Genomic Map