siRNA Details
| Gene Symbol: Smok2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191619.1 | NP_001178548.1 | 454 | |||
| XM_574319.4 | XP_574319.2 | 1 | 564 | |||
| Gene Symbol: LOC103689976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758866.1 | XP_008757088.1 | 1 | 466 | ||

Smok2a
Genomic Map