siRNA Details
| Gene Symbol: Smok2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191619.1 | NP_001178548.1 | 329 | |||
| XM_574319.4 | XP_574319.2 | 1 | 439 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smok2a| Gene Symbol: Smok2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191619.1 | NP_001178548.1 | 329 | |||
| XM_574319.4 | XP_574319.2 | 1 | 439 | |||