siRNA Details
| Gene Symbol: Ermard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728660.4 | XP_002728706.1 | 12 | 1158 | ||
| XM_006227972.2 | XP_006228034.1 | 12 | 1158 | |||
| XM_006227974.1 | XP_006228036.1 | 12 | 1263 | |||
| XM_006227975.2 | XP_006228037.1 | 12 | 1100 | |||
| XM_006227976.1 | XP_006228038.1 | 10 | 2393 | |||
| XM_008758815.1 | XP_008757037.1 | 10 | 871 | |||
| XM_008758816.1 | XP_008757038.1 | 8 | 705 | |||
| XM_008758817.1 | XP_008757039.1 | 8 | 643 | |||

Ermard
Genomic Map