siRNA Details
Gene Symbol: Ermard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728660.4 | XP_002728706.1 | 13 | 1342 | ||
XM_006227972.2 | XP_006228034.1 | 13 | 1342 | |||
XM_006227974.1 | XP_006228036.1 | 13 | 1447 | |||
XM_006227975.2 | XP_006228037.1 | 13 | 1284 | |||
XM_006227976.1 | XP_006228038.1 | 11 | 2577 | |||
XM_008758815.1 | XP_008757037.1 | 11 | 1055 | |||
XM_008758816.1 | XP_008757038.1 | 9 | 889 | |||
XM_008758817.1 | XP_008757039.1 | 9 | 827 |
