siRNA Details
| Gene Symbol: Lilrb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228238.2 | XP_006228300.1 | 1 | 40 | ||
| XM_006228239.2 | XP_006228301.1 | 1 | 40 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lilrb4| Gene Symbol: Lilrb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228238.2 | XP_006228300.1 | 1 | 40 | ||
| XM_006228239.2 | XP_006228301.1 | 1 | 40 | |||