siRNA Details
| Gene Symbol: Srp72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170601.1 | NP_001164072.1 | 1816 | |||
| XM_008770133.1 | XP_008768355.1 | 2262 | ||||
| GenBank mRNA | BC088335.1 | - | 1 | 72 | ||
| BC107914.1 | - | 966 | ||||
| BC158784.1 | - | 465 | 7302022 | |||

Srp72
Genomic Map