siRNA Details
| Gene Symbol: Mypop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109139.1 | NP_001102609.1 | 2, 3 | 670 | ||
| XM_006228452.2 | XP_006228514.1 | 2, 3 | 1669 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mypop| Gene Symbol: Mypop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109139.1 | NP_001102609.1 | 2, 3 | 670 | ||
| XM_006228452.2 | XP_006228514.1 | 2, 3 | 1669 | |||