siRNA Details
| Gene Symbol: Zfp94 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037212.2 | NP_001032289.2 | 5 | 705 | ||
| XM_006228456.2 | XP_006228518.1 | 5 | 1392 | |||
| XM_008758964.1 | XP_008757186.1 | 5 | 1321 | |||
| XM_008758965.1 | XP_008757187.1 | 5 | 1356 | |||
| XM_008758966.1 | XP_008757188.1 | 5 | 851 | |||
| XM_008758967.1 | XP_008757189.1 | 2 | 432 | |||
| GenBank mRNA | BC108299.1 | - | 5 | 705 | 7456297 | |

Zfp94
Genomic Map