siRNA Details
| Gene Symbol: Zfp420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728737.4 | XP_002728783.2 | 3 | 1211 | ||
| Gene Symbol: LOC103690163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759224.1 | XP_008757446.1 | 3 | 1211 | ||
| GenBank mRNA | FQ224550.1 | - | 3 | 1139 | ||

Zfp420
Genomic Map