siRNA Details
| Gene Symbol: Spib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024286.1 | NP_001019457.1 | 2, 3 | 53 | ||
| XM_008759404.1 | XP_008757626.1 | 1, 2 | 750 | |||
| GenBank mRNA | BC089881.1 | - | 2, 3 | 53 | 7368473 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spib| Gene Symbol: Spib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024286.1 | NP_001019457.1 | 2, 3 | 53 | ||
| XM_008759404.1 | XP_008757626.1 | 1, 2 | 750 | |||
| GenBank mRNA | BC089881.1 | - | 2, 3 | 53 | 7368473 | |