siRNA Details
Gene Symbol: Izumo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017514.1 | NP_001017514.1 | 2 | 449 | ||
XM_008759406.1 | XP_008757628.1 | 1 | 591 | |||
GenBank mRNA | AB195683.1 | - | 1 | 213 | ||
BC079244.1 | - | 2 | 449 | 7132791 |
