siRNA Details
| Gene Symbol: Gas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127504.1 | NP_001120976.1 | 5, 6 | 716 | ||
| XM_006229270.2 | XP_006229332.1 | 6, 7 | 1181 | |||
| XM_006229271.2 | XP_006229333.1 | 5, 6 | 635 | |||
| XM_006229273.2 | XP_006229335.1 | 6, 7 | 973 | |||
| XM_006229274.2 | XP_006229336.1 | 6, 7 | 1500 | |||
| XM_008759407.1 | XP_008757629.1 | 5, 6 | 714 | |||
| XM_008759408.1 | XP_008757630.1 | 5, 6 | 635 | |||

Gas2
Genomic Map