siRNA Details
| Gene Symbol: Wdr93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191949.1 | NP_001178878.1 | 1 | 103 | ||
| XM_006229401.2 | XP_006229463.1 | 2 | 294 | |||
| XM_006229403.2 | XP_006229465.1 | 2 | 239 | |||
| XM_006229404.2 | XP_006229466.1 | 2 | 294 | |||
| XM_008759547.1 | XP_008757769.1 | 2 | 2094 | |||
| XM_008759548.1 | XP_008757770.1 | 2 | 293 | |||
| XM_008759549.1 | XP_008757771.1 | 2 | 243 | |||
| XM_008759550.1 | XP_008757772.1 | 2 | 293 | |||
| XM_008759551.1 | XP_008757773.1 | 2 | 293 | |||
| XM_008759552.1 | XP_008757774.1 | 2 | 293 | |||
| XM_008759553.1 | XP_008757775.1 | 2 | 292 | |||

Wdr93
Genomic Map