siRNA Details
| Gene Symbol: Ddias | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126294.1 | NP_001119766.1 | 5 | 459 | ||
| GenBank mRNA | BC166470.1 | - | 5 | 459 | 9088340 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddias| Gene Symbol: Ddias | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126294.1 | NP_001119766.1 | 5 | 459 | ||
| GenBank mRNA | BC166470.1 | - | 5 | 459 | 9088340 | |