siRNA Details
| Gene Symbol: LOC499235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_574528.5 | XP_574528.1 | 1, 2 | 331 | ||
| GenBank mRNA | AY539892.1 | - | 1, 2 | 331 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499235| Gene Symbol: LOC499235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_574528.5 | XP_574528.1 | 1, 2 | 331 | ||
| GenBank mRNA | AY539892.1 | - | 1, 2 | 331 | ||