siRNA Details
| Gene Symbol: LOC499240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024287.1 | NP_001019458.1 | 5, 6 | 688 | ||
| GenBank mRNA | BC087685.1 | - | 5, 6 | 688 | 7309316 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499240| Gene Symbol: LOC499240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024287.1 | NP_001019458.1 | 5, 6 | 688 | ||
| GenBank mRNA | BC087685.1 | - | 5, 6 | 688 | 7309316 | |