siRNA Details
| Gene Symbol: Smg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759828.1 | XP_008758050.1 | 24 | 3992 | ||
| GenBank mRNA | FQ226191.1 | - | 1 | 122 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smg1| Gene Symbol: Smg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759828.1 | XP_008758050.1 | 24 | 3992 | ||
| GenBank mRNA | FQ226191.1 | - | 1 | 122 | ||