siRNA Details
| Gene Symbol: Itpripl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127303.1 | NP_001120775.1 | 1 | 1701 | ||
| GenBank mRNA | BC166746.1 | - | 1 | 1701 | 5598710 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itpripl2| Gene Symbol: Itpripl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127303.1 | NP_001120775.1 | 1 | 1701 | ||
| GenBank mRNA | BC166746.1 | - | 1 | 1701 | 5598710 | |