siRNA Details
| Gene Symbol: Knop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134611.2 | NP_001128083.1 | 2 | 628 | ||
| XM_008759787.1 | XP_008758009.1 | 2 | 636 | |||
| XM_008759788.1 | XP_008758010.1 | 2 | 636 | |||
| XM_008759789.1 | XP_008758011.1 | 2 | 769 | |||

Knop1
Genomic Map