siRNA Details
| Gene Symbol: Apobr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109154.1 | NP_001102624.1 | 3 | 1780 | ||
| XM_006230319.2 | XP_006230381.1 | 2 | 2439 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apobr| Gene Symbol: Apobr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109154.1 | NP_001102624.1 | 3 | 1780 | ||
| XM_006230319.2 | XP_006230381.1 | 2 | 2439 | |||