siRNA Details
| Gene Symbol: Itgax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230382.2 | XP_006230444.1 | 22 | 3050 | ||
| XM_574569.6 | XP_574569.3 | 26 | 3374 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itgax| Gene Symbol: Itgax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230382.2 | XP_006230444.1 | 22 | 3050 | ||
| XM_574569.6 | XP_574569.3 | 26 | 3374 | |||