siRNA Details
| Gene Symbol: LOC499276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230423.2 | XP_006230485.1 | 3 | 575 | ||
| GenBank mRNA | BC098034.1 | - | 3 | 535 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499276| Gene Symbol: LOC499276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230423.2 | XP_006230485.1 | 3 | 575 | ||
| GenBank mRNA | BC098034.1 | - | 3 | 535 | ||