siRNA Details
| Gene Symbol: Tex36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024288.1 | NP_001019459.1 | 451 | |||
| GenBank mRNA | BC079265.1 | - | 451 | 7133623 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex36| Gene Symbol: Tex36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024288.1 | NP_001019459.1 | 451 | |||
| GenBank mRNA | BC079265.1 | - | 451 | 7133623 | ||