siRNA Details
| Gene Symbol: Ano9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230589.2 | XP_006230651.1 | 10 | 872 | ||
| XM_006230590.2 | XP_006230652.1 | 7 | 656 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ano9| Gene Symbol: Ano9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230589.2 | XP_006230651.1 | 10 | 872 | ||
| XM_006230590.2 | XP_006230652.1 | 7 | 656 | |||