siRNA Details
| Gene Symbol: Ptprcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024289.1 | NP_001019460.1 | 2 | 373 | ||
| GenBank mRNA | BC088279.1 | - | 2 | 373 | 7382668 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprcap| Gene Symbol: Ptprcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024289.1 | NP_001019460.1 | 2 | 373 | ||
| GenBank mRNA | BC088279.1 | - | 2 | 373 | 7382668 | |