siRNA Details
| Gene Symbol: Gal3st3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024290.1 | NP_001019461.1 | 3 | 702 | ||
| GenBank mRNA | BC094516.1 | - | 3 | 702 | 7314624 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gal3st3| Gene Symbol: Gal3st3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024290.1 | NP_001019461.1 | 3 | 702 | ||
| GenBank mRNA | BC094516.1 | - | 3 | 702 | 7314624 | |