siRNA Details
| Gene Symbol: LOC499306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024291.1 | NP_001019462.1 | 5 | 456 | ||
| GenBank mRNA | BC078985.1 | - | 5 | 456 | 7112339 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499306| Gene Symbol: LOC499306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024291.1 | NP_001019462.1 | 5 | 456 | ||
| GenBank mRNA | BC078985.1 | - | 5 | 456 | 7112339 | |