siRNA Details
| Gene Symbol: Zbtb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109162.2 | NP_001102632.1 | 2 | 435 | ||
| XM_006230992.2 | XP_006231054.1 | 2 | 353 | |||
| GenBank mRNA | BC160822.1 | - | 2 | 435 | 7380153 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb3| Gene Symbol: Zbtb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109162.2 | NP_001102632.1 | 2 | 435 | ||
| XM_006230992.2 | XP_006231054.1 | 2 | 353 | |||
| GenBank mRNA | BC160822.1 | - | 2 | 435 | 7380153 | |