siRNA Details
| Gene Symbol: Nmrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024292.1 | NP_001019463.1 | 3 | 184 | ||
| XM_008760290.1 | XP_008758512.1 | 3 | 316 | |||
| GenBank mRNA | BC079144.1 | - | 3 | 184 | 7128209 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nmrk1| Gene Symbol: Nmrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024292.1 | NP_001019463.1 | 3 | 184 | ||
| XM_008760290.1 | XP_008758512.1 | 3 | 316 | |||
| GenBank mRNA | BC079144.1 | - | 3 | 184 | 7128209 | |