siRNA Details
| Gene Symbol: RGD1560242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109164.1 | NP_001102634.1 | 6 | 613 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1560242| Gene Symbol: RGD1560242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109164.1 | NP_001102634.1 | 6 | 613 | ||