siRNA Details
| Gene Symbol: Fam178a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134612.1 | NP_001128084.1 | 9, 10 | 2497 | ||
| XM_006231550.2 | XP_006231612.1 | 8, 9 | 2486 | |||
| XM_006231551.2 | XP_006231613.1 | 8, 9 | 2485 | |||

Fam178a
Genomic Map