siRNA Details
| Gene Symbol: C1H10orf76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_574677.4 | XP_574677.2 | 13 | 1124 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1H10orf76| Gene Symbol: C1H10orf76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_574677.4 | XP_574677.2 | 13 | 1124 | ||