siRNA Details
| Gene Symbol: Clasrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024294.1 | NP_001019465.2 | 16, 17 | 1813 | ||
| XM_008758970.1 | XP_008757192.1 | 16, 17 | 1884 | |||
| XM_008758971.1 | XP_008757193.1 | 16, 17 | 1798 | |||
| GenBank mRNA | BC089090.1 | - | 16, 17 | 1813 | 7111206 | |

Clasrp
Genomic Map