siRNA Details
| Gene Symbol: Sumo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024295.1 | NP_001019466.1 | 2 | 126 | ||
| GenBank mRNA | BC083728.1 | - | 2 | 126 | 7191303 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sumo3| Gene Symbol: Sumo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024295.1 | NP_001019466.1 | 2 | 126 | ||
| GenBank mRNA | BC083728.1 | - | 2 | 126 | 7191303 | |