siRNA Details
Gene Symbol: Spatc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024296.1 | NP_001019467.1 | 4 | 959 | ||
GenBank mRNA | BC079291.1 | - | 4 | 959 | 7134390 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spatc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024296.1 | NP_001019467.1 | 4 | 959 | ||
GenBank mRNA | BC079291.1 | - | 4 | 959 | 7134390 |