siRNA Details
| Gene Symbol: Pkm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243187.2 | XP_006243249.1 | 11 | 2414 | ||
| XM_006243188.2 | XP_006243250.1 | 11 | 2414 | |||
| XM_006243190.2 | XP_006243252.1 | 12 | 2486 | |||
| GenBank mRNA | AY724474.1 | - | 11 | 2181 | ||
| BC061541.1 | - | 11 | 2059 | 6888187 | ||
| FQ223721.1 | - | 11 | 2192 | |||
| FQ229350.1 | - | 2 | 784 | |||
| Gene Symbol: LOC100362738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772937.1 | XP_008771159.1 | 2 | 2151 | ||

Pkm
Genomic Map