siRNA Details
| Gene Symbol: Zfp365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025145.1 | NP_001020316.1 | 3 | 1094 | ||
| GenBank mRNA | BC087056.1 | - | 3 | 1094 | 7124000 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp365| Gene Symbol: Zfp365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025145.1 | NP_001020316.1 | 3 | 1094 | ||
| GenBank mRNA | BC087056.1 | - | 3 | 1094 | 7124000 | |