siRNA Details
| Gene Symbol: Lrrc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109171.1 | NP_001102641.1 | 5 | 596 | ||
| XM_006256465.2 | XP_006256527.1 | 5 | 622 | |||
| XM_006256466.2 | XP_006256528.1 | 5 | 1414 | |||
| XM_006256467.2 | XP_006256529.1 | 5 | 643 | |||
| XM_006256468.2 | XP_006256530.1 | 5 | 655 | |||
| XM_008772902.1 | XP_008771124.1 | 4 | 418 | |||
| XM_008772903.1 | XP_008771125.1 | 4 | 1385 | |||
| GenBank mRNA | FQ223562.1 | - | 5 | 590 | ||

Lrrc20
Genomic Map