siRNA Details
| Gene Symbol: Lrrc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109171.1 | NP_001102641.1 | 4 | 524 | ||
| XM_006256465.2 | XP_006256527.1 | 4 | 547 | |||
| XM_006256466.2 | XP_006256528.1 | 4 | 1339 | |||
| XM_006256467.2 | XP_006256529.1 | 4 | 568 | |||
| XM_006256468.2 | XP_006256530.1 | 4 | 580 | |||
| XM_008772902.1 | XP_008771124.1 | 3 | 346 | |||
| XM_008772903.1 | XP_008771125.1 | 3 | 1313 | |||
| GenBank mRNA | FQ223562.1 | - | 4 | 518 | ||

Lrrc20
Genomic Map