siRNA Details
| Gene Symbol: Col13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109172.1 | NP_001102642.1 | 41 | 2939 | ||
| XM_006256479.2 | XP_006256541.1 | 41 | 2974 | |||
| XM_006256481.2 | XP_006256543.1 | 37 | 2817 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col13a1| Gene Symbol: Col13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109172.1 | NP_001102642.1 | 41 | 2939 | ||
| XM_006256479.2 | XP_006256541.1 | 41 | 2974 | |||
| XM_006256481.2 | XP_006256543.1 | 37 | 2817 | |||