siRNA Details
| Gene Symbol: Lims1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145456.1 | NP_001138928.1 | 2 | 317 | ||
| XM_006256385.2 | XP_006256447.1 | 2 | 2219 | |||
| XM_006256386.2 | XP_006256448.1 | 2 | 2219 | |||
| XM_006256387.2 | XP_006256449.1 | 2 | 2216 | |||
| XM_006256388.2 | XP_006256450.1 | 2 | 2083 | |||
| XM_006256389.2 | XP_006256451.1 | 2 | 310 | |||
| XM_006256390.2 | XP_006256452.1 | 2 | 1890 | |||
| XM_006256391.2 | XP_006256453.1 | 2 | 1889 | |||
| GenBank mRNA | FJ640984.1 | - | 2 | 317 | ||
| FJ640985.1 | - | 2 | 396 | |||
| FQ222665.1 | - | 2 | 405 | |||

Lims1
Genomic Map